Tweezers with Delrin tips
Lieferant: OUTILS RUBIS
49D
OUTI49DEA 82 EUR
OUTI49D
Tweezers with Delrin tips
Pinzetten
Stainless steel tweezers with Delrin tips. Excellent for use in Biotech applications.
  • Temperature resistant up to 115 °C
  • Very soft material, non scratching, nicking or marring