AEROSIL® R 805

Lieferant: OVERLACK CHEMISCHE PRODUKTE

AEROSILR805
OVERAEROSILR805EA 804 EUR
OVERAEROSILR805
AEROSIL® R 805
AEROSIL®
MDL: MFCD00011232
CAS-Nummer: 112945-52-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR