AEROSIL® R 812S

Lieferant: OVERLACK CHEMISCHE PRODUKTE

AEROSILR812S
OVERAEROSILR812SEA 649 EUR
OVERAEROSILR812S
AEROSIL® R 812S
AEROSIL®
MDL: MFCD00011232
CAS-Nummer: 112945-52-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR