Vitamin B12 (Cyanocobalamin)
Lieferant: Oxford Biotechnology
CBL64
CBL64
CHMCCBL64EA
0
EUR
CHMCCBL64
OXBTCBL64
Vitamin B12 (Cyanocobalamin)
Vitamin B12 (Cyanocobalamin)
Formel:
C₆₃H₈₈CoN₁₄O₁₄P Molecular Weight: 1355.38 g/mol Schmelzpunkt: 320 Lagertemperatur: Kühlschrank |
MDL:
MFCD00151092 CAS-Nummer: 68-19-9 EINECS: 200-680-0 Merck Index: 12,10152 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...