Flow Cell Priming Kit
Lieferant: Oxford Nanopore Technologies
Additional flow cell priming reagents to enable additional loading of more library during a sequencing run.
- The kit can be used to prime a MinION™ Flow Cell for sequencing or to add more fuel to a flow cell during a sequencing experiment.
Flow Cell Priming Kit V14 (EXP-FLP004) ― the Flow Cell Priming Kit V14 contains Flow Cell Flush (FCF) and Flow Cell Tether (FCT), sufficient for 6 reactions. This kit is compatible with the latest library preparation kit chemistry (V14) and R10.4.1 Flow Cells.
Note: This kit is not compatible with the Ultra-Long DNA Sequencing Kit V14, which requires specific flow cell priming reagents.
Lieferumfang: Kits are shipped at 2 to 8 °C in an environmentally-friendly temperature-controlled shipping box. Upon receipt, please place the product in a long-term storage location at −20 °C. Oxford Nanopore Technologies deem the useful life of the product to be 3 months from receipt by the customer.
Bitte beachten Oxford Nanopore Technologies products are not intended for use for health assessment or to diagnose, treat, mitigate, cure, or prevent any disease or condition.