Long Fragment Buffer Expansion
Lieferant: Oxford Nanopore Technologies
Additional Long Fragment Buffer (LFB).
The LFB expansion contains additional Long Fragment Buffer (LFB) for use with sequencing experiments where shorter fragments (<3 kb) are to be removed. The LFB expansion contains enough reagents for four reactions.
Lieferumfang: Kits are shipped at 2 to 8 °C in an environmentally-friendly temperature-controlled shipping box. Upon receipt, please place the product in a long-term storage location at −20 °C. Oxford Nanopore Technologies deem the useful life of the product to be 3 months from receipt by the customer.