2,3,5-Triphenyltetrazoliumchlorid 0,125% in wässriger Lösung

Lieferant: THERMO FISHER DIAGNOSTICS

Synonyme: 2,3,5-Triphenyl-2H-tetrazol Hydrochlorid, TTC, Tetrazoliumrotchlorid

SR0148A
OXOISR0148AEA 93.6 EUR
OXOISR0148A
2,3,5-Triphenyltetrazoliumchlorid 0,125% in wässriger Lösung
2,3,5-Triphenyltetrazoliumchlorid
Formel: C₁₉H₁₅ClN₄
Molecular Weight: 334,81 g/mol
MDL: MFCD00011963
CAS-Nummer: 298-96-4
EINECS: 206-071-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR