Transfection boosters, GeneBlaster™
Lieferant: OZ BIOSCIENCES
GB20010
GB20011
GB20012
GB20013
GB20014
OZBIGB20010EA
162.95
EUR
OZBIGB20010
OZBIGB20011
OZBIGB20012
OZBIGB20013
OZBIGB20014
Transfection boosters, GeneBlaster™
Transfektionsreagenzien
GeneBlaster™ transfection boosters are an innovative and efficient solution to improving gene expression levels. GeneBlaster™ kits are a set of chemicals designed to get higher and longer transgene expression levels.
- Higher gene expression in many cells
- Convenient for a large panel of adherent and suspension cells
- Prolong in vitro gene expression
- Successful with all genetic vectors
- Simple, rapid, and easy to use
Store all GeneBlaster™ kits at +4 °C, except GeneBlaster™ Emerald at –20 °C.
Lieferumfang: Shipped at room temperature.