siRNA transfection reagent, SilenceMag™
Lieferant: OZ BIOSCIENCES
SM10200
SM10500
SM11000
SM13000
KC30300
KC30400
OZBISM10200EA
117.18
EUR
OZBISM10200
OZBISM10500
OZBISM11000
OZBISM13000
OZBIKC30300
OZBIKC30400
siRNA transfection reagent, SilenceMag™
Transfektionsreagenzien
SilenceMag™ siRNA delivery reagent is based on Magnetofection™ technology. Specifically designed for siRNA transfection, SilenceMag™ gives reliable high protein knockdown at very low doses of siRNA in numerous cell types (primary cells, hard-to-transfect and cell lines).
- High protein knockdown efficiency
- For all siRNA applications
- No off-target effects
- Serum compatible and non toxic
- Simple, rapid, and easy to use
Store at +4 °C.
Zubehörinformationen: Needs to be used with a magnetic plate, available as a separate accessory or as part of a starter kit.
Lieferumfang: Shipped at room temperature.