Transfection reagent for viral capture, concentration and storage, Mag4C-LV
Lieferant: OZ BIOSCIENCES
LTK11200 LKC11000
OZBILTK11200EA 177.86 EUR
OZBILTK11200 OZBILKC11000
Transfection reagent for viral capture, concentration and storage, Mag4C-LV
Transfektionsreagenzien
Mag4C-LV is designed for magnetic virus concentration. This kit is specifically developed for capturing, concentrating and storing lentiviruses and retroviruses.
  • Simple, rapid and ready to use
  • Concentration of lentiviruses by magnetic capture in 30 to 45 minutes
  • High yield of viral capture and recovery
  • No need to process magnetic beads before capture
  • Avoid ultracentrifugation, precipitation and chemicals, reduced handling steps of viruses
  • Improve virus stability under storage conditions
Mag4C-LV magnetic nanoparticles capture viruses in culture media with 80 to 99% efficiency. Once captured onto magnetic beads, viruses can be:
Concentrated and stored with the storage buffer or directly used for downstream assay.
Concentrated, eluted from the magnetic beads with the elution buffer and stored with the storage buffer or used for subsequent assays.
Zubehörinformationen: Multipurpose magnetic separation rack for 50, 15 or 1,5 ml tubes needs to be used with the kit (available separately).
Bitte beachten Store at +4 °C.