R-Phycoerythrin
Lieferant: OZ BIOSCIENCES
It is a naturally fluorescent protein from algae and is composed of a 240 kD protein covalently linked to a number of fluorophores called phycobilins. Its exceptional fluorescence characteristics make it a perfect positive control for intracellular protein delivery using OZ Biosciences Pro-DeliverIN reagent. R-phycoerythrin is provided at a 100 μg/ml concentration in DPBS pH 7,4.
- Absorption maximum: 498/565 nm
- Emission maximum: 573 nm
- Extinction coefficient: 1,96×10⁶ M⁻¹ cm⁻¹
- Quantum yield - 0,84
Store at +4 °C and protect from the light.
Application: Transfection control
Lieferumfang: Shipped at room temperature.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...