Stem transfection reagents, DreamFect™ Stem
Lieferant: OZ BIOSCIENCES
ST30500 ST31000
OZBIST30500EA 232.17 EUR
OZBIST30500 OZBIST31000
Stem transfection reagents, DreamFect™ Stem
Transfektionsreagenzien
DreamFect™ Stem is a powerful transfection reagent allowing multipotent stem cell transfection with high efficiency and very low toxicity. Its specific composition based on TEE-technology allows transfecting embryonic and multipotent stem cells in the presence of serum, maintaining their undifferentiated stage and their capacity to differentiate.
  • High transfection efficiency for multipotent stem cells
  • Minimised toxicity due to reagent biodegradability and low DNA amount required
  • Cell phenotype and differentiation potential are not affected
  • Serum compatible
  • Simple, ready to use and rapid
Store at –20 °C.
Lieferumfang: Shipped at room temperature.