Cell culture media, Dulbecco's Modified Eagle Medium (DMEM)
Lieferant: Biowest
P0103-N10L P0061-N1L P0061-N10L P0061-N5L P0102-N1L P0102-N10L P0102-N5L P0103-N1L L0060-500 L0064-500 L0066-500 L0101-500 L0102-500 L0103-500 L0100-500 L0104-500 L0106-500 L0107-500 P0102-N50L P0103-N5L P0061-N50L L0065-500 P0103-N50L
P0103-N10LEA 145 EUR
P0103-N10L P0061-N1L P0061-N10L P0061-N5L P0102-N1L P0102-N10L P0102-N5L P0103-N1L L0060-500 L0064-500 L0066-500 L0101-500 L0102-500 L0103-500 L0100-500 L0104-500 L0106-500 L0107-500 P0102-N50L P0103-N5L P0061-N50L L0065-500 P0103-N50L
Cell culture media, Dulbecco's Modified Eagle Medium (DMEM)
Zellkulturmedien
Dulbecco's Modified Eagle Medium (DMEM) is a modification of Basal Medium Eagle (BME) which contains approximately four times as much of the vitamins and amino acids, and two to four times as much glucose (depending on the version) as the original formulation.
  • Suitable for most cell types
  • Available in a broad variety of formulations
  • Also available with stable glutamine (L-Alanyl-L-Glutamine) for a better shelf life