Cell culture media, Dulbecco's Modified Eagle Medium (DMEM)
Lieferant: Biowest
P0103-N10L
P0061-N1L
P0061-N10L
P0061-N5L
P0102-N1L
P0102-N10L
P0102-N5L
P0103-N1L
L0060-500
L0064-500
L0066-500
L0101-500
L0102-500
L0103-500
L0100-500
L0104-500
L0106-500
L0107-500
P0102-N50L
P0103-N5L
P0061-N50L
L0065-500
P0103-N50L
P0103-N10LEA
145
EUR
P0103-N10L
P0061-N1L
P0061-N10L
P0061-N5L
P0102-N1L
P0102-N10L
P0102-N5L
P0103-N1L
L0060-500
L0064-500
L0066-500
L0101-500
L0102-500
L0103-500
L0100-500
L0104-500
L0106-500
L0107-500
P0102-N50L
P0103-N5L
P0061-N50L
L0065-500
P0103-N50L
Cell culture media, Dulbecco's Modified Eagle Medium (DMEM)
Zellkulturmedien
Dulbecco's Modified Eagle Medium (DMEM) is a modification of Basal Medium Eagle (BME) which contains approximately four times as much of the vitamins and amino acids, and two to four times as much glucose (depending on the version) as the original formulation.
- Suitable for most cell types
- Available in a broad variety of formulations
- Also available with stable glutamine (L-Alanyl-L-Glutamine) for a better shelf life