Silbernitrat 99,0-100,5 Ph. Eur., Macron Fine Chemicals™

Lieferant: Avantor Performance Materials

Synonyme: Höllenstein

P100-31EA 0 EUR
P100-31 P100-33 P100-36 P100-42 MACRP100-38
Silbernitrat 99,0-100,5 Ph. Eur., Macron Fine Chemicals™
Silbernitrat
Formel: AgNO₃
Molecular Weight: 169.87 g/mol
Siedepunkt: 444
Schmelzpunkt: 210
MDL: MFCD00003414
CAS-Nummer: 7761-88-8
UN: 1493
ADR: 5.1 II
Merck Index: 12,08661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Ph. Eur. - Assay 99.0 - 100.5%
Ph. Eur. - Identification A Passes Test
Ph. Eur. - Identification B Passes Test
Ph. Eur. - Appearance of Solution Passes Test
Ph. Eur. - Acidity or Alkalinity Passes Test
Ph. Eur. - Foreign Salts ≤0.3%
Ph. Eur. - Aluminium, Lead, Copper and Bismuth Passes Test
Clarity and Colour of solution Clear, Colourless
Identity A Positive Reaction
Identity B Positive Reaction
Copper Does not Appear Blue Colour
Assay 99.8 - 100.5%
For Manufacturing, Processing or ph. Eur. Repackaging
Bulk Pharmaceutical Chemical
No Class 1,2,3 or Other Solvents Are Used Or Produced In the Manufacturing or Purification of the Product
Meets Ph. Eur. Requirements


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR