Citronensäure Monohydrat Ph. Eur., E330, Macron Fine Chemicals™
Lieferant: Avantor Performance Materials
P101-33EA
0
EUR
P101-33
P101-38
Citronensäure Monohydrat Ph. Eur., E330, Macron Fine Chemicals™
Formel:
C₆H₈O₇.H₂O |
MDL:
MFCD00149972 CAS-Nummer: 5949-29-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...