Calciumcarbonat Ph. Eur., Macron Fine Chemicals™

Lieferant: Avantor Performance Materials

P104-33EA 45.6 EUR
P104-33 P104-38
Calciumcarbonat Ph. Eur., Macron Fine Chemicals™
Calciumcarbonat
Formel: CaCO₃
MDL: MFCD00010906
CAS-Nummer: 471-34-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR