tri-Natriumcitrat Dihydrat, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials

P105-38EA 0 EUR
P105-38
tri-Natriumcitrat Dihydrat, Macron Fine Chemicals™
tri-Natriumcitrat Dihydrat
Formel: Na₃C₆H₅O₇·2H₂O
MDL: MFCD00150031
CAS-Nummer: 6132-04-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e32147a215e8895f4b6db5aab1b99da667b795ac&vid=b929cda9d6f2eed18a8fefe57aa02b561af7c97f&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR