Natriumdihydrogenphosphat Dihydrat Ph. Eur., Macron Fine Chemicals™
Lieferant: Avantor Performance Materials
Synonyme:
Natriumphosphat monobasisch Dihydrat
P108-42EA
0
EUR
P108-42
Natriumdihydrogenphosphat Dihydrat Ph. Eur., Macron Fine Chemicals™
Natriumdihydrogenphosphat Dihydrat
Formel:
NaH₂PO₄·2H₂O Molecular Weight: 156,01 g/mol |
MDL:
MFCD00149209 CAS-Nummer: 13472-35-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...