Salzsäure Ph. Eur., Macron Fine Chemicals™

Lieferant: Avantor Performance Materials

P112-33EA 30.79 EUR
P112-33 P112-36
Salzsäure Ph. Eur., Macron Fine Chemicals™
Salzsäure
Formel: HCl
MDL: MFCD00011324
CAS-Nummer: 7647-01-0
UN: 1789
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR