Aceton 99,0 Ph. Eur., Macron Fine Chemicals™
Lieferant: Avantor Performance Materials
Synonyme:
Propanon, DMK, 2-Propanon, Dimethylketon
P113-33EA
65.99
EUR
P113-33
P113-38
P113-39
P113-40
Aceton 99,0 Ph. Eur., Macron Fine Chemicals™
Aceton
Formel:
CH₃COCH₃ Molecular Weight: 58.08 g/mol Siedepunkt: 56.2 Schmelzpunkt: -95.4 |
MDL:
MFCD00008765 CAS-Nummer: 67-64-1 UN: 1090 ADR: 3 II Merck Index: 13,00067 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...