Aceton ≥99,0% Ph. Eur., Macron Fine Chemicals™

Lieferant: Avantor Performance Materials

Synonyme: Propanon, DMK, 2-Propanon, Dimethylketon

P113-33EA 65.99 EUR
P113-33 P113-38 P113-39 P113-40
Aceton ≥99,0% Ph. Eur., Macron Fine Chemicals™
Aceton
Formel: CH₃COCH₃
MDL: MFCD00008765
CAS-Nummer: 67-64-1
UN: 1090
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR