di-Natriumhydrogenphosphat Dodecahydrat Ph. Eur., Macron Fine Chemicals™

Lieferant: Avantor Performance Materials

P116-33EA 75.29 EUR
P116-33 P116-42
di-Natriumhydrogenphosphat Dodecahydrat Ph. Eur., Macron Fine Chemicals™
di-Natriumhydrogenphosphat Dodecahydrat
Formel: Na₂HPO₄·12H₂O
MDL: MFCD00149181
CAS-Nummer: 10039-32-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR