Natriumdihydrogenphosphat Monohydrat Ph. Eur., Macron Fine Chemicals™

Lieferant: Avantor Performance Materials

Synonyme: Natriumphosphat monobasisch Monohydrat

P117-38EA 1550 EUR
P117-38
Natriumdihydrogenphosphat Monohydrat Ph. Eur., Macron Fine Chemicals™
Natriumdihydrogenphosphat Monohydrat
Formel: NaH₂PO₄·H₂O
Molecular Weight: 137,99 g/mol
Schmelzpunkt: 100 °C
Dichte: 2,04 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00149208
CAS-Nummer: 10049-21-5
EINECS: 231-449-2
REACH: 01-2119489796-13
Merck Index: 13,08734
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4e3b73d9200cc18a9423c5b98ad4affbd4aca8a&vid=a46f80a2f4b075e731486341a7be9b01c444a160&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR