Aluminiumkaliumsulfat Dodecahydrat Ph. Eur., Macron Fine Chemicals™
Lieferant: Avantor
Synonyme:
Alumen, Kalialaun, Aluminiumkaliumbis(sulfat) Dodecahydrat
P124-38EA
0
EUR
P124-38
Aluminiumkaliumsulfat Dodecahydrat Ph. Eur., Macron Fine Chemicals™
Aluminiumkaliumsulfat Dodecahydrat
Formel:
AlK(SO₄)₂·12H₂O Molecular Weight: 474,39 g/mol |
MDL:
MFCD00149143 CAS-Nummer: 7784-24-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...