Natriumhydrogencarbonat Ph. Eur., Macron Fine Chemicals™
Lieferant: Avantor Performance Materials
P132-38EA
0
EUR
P132-38
Natriumhydrogencarbonat Ph. Eur., Macron Fine Chemicals™
Natriumhydrogencarbonat
Formel:
NaHCO₃ |
MDL:
MFCD00003528 CAS-Nummer: 144-55-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...