Calciumchlorid Hexahydrat Ph. Eur., Macron Fine Chemicals™
Lieferant: Avantor Performance Materials
|
Achtung
|
P133-36EA
432.85
EUR
P133-36
Calciumchlorid Hexahydrat Ph. Eur., Macron Fine Chemicals™
Calciumchlorid Hexahydrat
Formel:
CaCl₂·6H₂O Molecular Weight: 219,08 g/mol Schmelzpunkt: 29,54 °C Dichte: 1,71 g/cm³ (25 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD00149616 CAS-Nummer: 7774-34-7 EINECS: 233-140-8 Merck Index: 13,01660 |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...