Zinksulfat Heptahydrat Ph. Eur., Macron Fine Chemicals™

Lieferant: Avantor Performance Materials

P134-33EA 94.1 EUR
P134-33 P134-38
Zinksulfat Heptahydrat Ph. Eur., Macron Fine Chemicals™
Zinksulfat Heptahydrat

Standards: Meets Ph. Eur. chemical specifications.

Formel: ZnSO₄·7H₂O
MDL: MFCD00149894
CAS-Nummer: 7446-20-0
UN: 3077
ADR: 9,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR