Proflavine hemisulphate hydrate, powder, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyme: 3,6-Diaminoacridine hemisulphate salt, 3,6'-Diaminoacridine sulphate, 3,6-Diaminoacridinium monohydrogen sulphate

MQ100

P2508-10G P2508-25G
P2508-10GEA 61.1 EUR
P2508-10G P2508-25G
Proflavine hemisulphate hydrate, powder, Sigma-Aldrich®
Proflavine hemisulphate hydrate

Proflavine is a small, amphipathic, acridine-derived fluorescent dye.Proflavine hemisulfate is used as a rapid stain for cytological studies of biological specimens. It stains cytoplasmic structures and nuclei. Proflavine has the capability to intercalate double stranded DNA.


Proflavine hemisulfate salt hydrate has been used to stain cells of the 1483 cell line. It has also been used as a calibrating dye in fluorescence resonance energy transfer (FRET) analysis.

Formel: (C₁₃H₁₁N₃)₂·H₂SO₄·xH₂O
Molecular Weight: 516.58 g/mol (anhy)
Lagertemperatur: Room Temperature
MDL: MFCD00149650
CAS-Nummer: 1811-28-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=793e18138ad1dcd9bab9827c5b4ea0a650f11c67&vid=bc062d11ec5b30dfdf3bbea87deb6fd4787afa1e&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) Faint Orange to Red-Brown to Brown
Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Water (by Karl Fischer) ≤10.0 %
Solubility (Colour) Faint Orange to Dark Brown
Solubility (Form)
10 mg/ml, H2O
Solution
Carbon
Anhydrous
59.2 - 61.7 %
Nitrogen
Anhydrous
15.9 - 16.6 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR