Paraplast® für Gewebe, Einbettmedium
Lieferant: SIGMA ALDRICH MICROSCOPY
M-Clarity™ Qualitätsstufe = MQ200
P3558-1KG
P3558-1KGEA
51.3
EUR
P3558-1KG
Paraplast® für Gewebe, Einbettmedium
Paraffinwachs versetzt mit synthetischen Polymeren
Paraplast® is a paraffin–polyisobutylene mixture for tissue embedding and infiltration. It is a refined combination of highly purified paraffin with plastic polymers. It produces minimal tissue compression and wrinkle-free sections and cuts to 4 μm thickness with excellent ribbon continuity.
- Paraplast® has been used for tissue embedding and in the following applications:
- To screen major groups of vertebrates for the presence of cardiac smooth muscle and its phylogenetic distribution in terrestrial and aquatic turtles
- To study the relationship between membrane impedance and dendrite length of hippocampal neurons
- To compare the regenerative properties of human stem cells of the apical papilla as an alternative to other dental stem cells
Minimal tissue compression. Produces wrinkle-free sections. Excellent ribbon continuity. Double filtered paraffin wax in pellet form.
Schmelzpunkt:
56,..57 °C (Lit.) Lagertemperatur: Room Temperature |
CAS-Nummer:
8002-74-2 |
Spezifikation Testergebnisse
Note | Paraplast is a registered trademark of Leica Biosystems |
Moisture content < or = 0.2% |
Conforms |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...