Paraplast Plus® für Gewebe, Einbettmedium

Lieferant: SIGMA ALDRICH MICROSCOPY


M-Clarity™ Qualitätsstufe = MQ100

P3683-1KG
P3683-1KGEA 57.7 EUR
P3683-1KG
Paraplast Plus® für Gewebe, Einbettmedium
Paraffinwachs versetzt mit synthetischen Polymeren

Paraplast Plus® is a tissue embedding medium comprising highly purified paraffin and polyisobutylene with approximately 0.8% DMSO added to improve infiltration, sectioning, and penetration into tissues. It is recommended for tissues that are difficult to process with regular Paraplast®. Paraplast Plus® cuts continuous ribbons at 2 μm thickness or short ribbons at 1 μm thickness.


  • Paraplast Plus® has been used in the examination of vertebral specimens of Malacoraja senta
  • It has also been used in the identification and expression analysis of PIN genes in rice
  • It has been used in the study of glucose transport mechanisms in uterine tissues during pregnancy
  • It has been used to embed Bouin's fixed carp tissues, rat ventral prostatic fragments, and common bean (Phaseolus vulgaris L.) nodules

Schmelzpunkt: 56,..57 °C (Lit.)
Lagertemperatur: Room Temperature
CAS-Nummer: 8002-74-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR