Amphotericin B

Lieferant: Biowest

Synonyme: (1R,3S,5R,6R,9R,11R,15S,16R,17R,18S,19E,21E,23E,25E,27E,29E,31E,33R,35S,36R,37S)-33-(((2R,3S,4S,5S,6R)-4-Amino-3,5-dihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-1,3,5,6,9,11,17,37-octahydroxy-15,16,18-trimethyl-13-oxo-14,39-dioxabicyclo[33.3.1]nonatriaconta-19,21,23,25,27,29,31-heptaene-36-carboxylic acid

P4030-250MG
P4030-250MGEA 52.5 EUR
P4030-250MG
Amphotericin B
Amphotericin B

Amphotericin B is used for the prevention or elimination of fungal contaminants in cell culture.


  • Yellow to orange powder
  • Storage temperature of 2 to 8 °C
  • Shelf life of 24 months


Interferes with fungal membrane permeability by forming channels in the membranes and causing small molecules to leak out.


Bitte beachten This product is intended to be used in vitro, in laboratory only. Not recommended to be used in therapy, human or veterinary applications.

Formel: C₄₇H₇₃NO₁₇
Molecular Weight: 924,08 g/mol
MDL: MFCD00877763
CAS-Nummer: 1397-89-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR