Phloxin B, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: CI 45410

P4030-25G
P4030-25GEA 336 EUR
P4030-25G
Phloxin B, Sigma-Aldrich®
Phloxin B

Phloxine B has been used as a component of yeast extract peptone dextrose (YEPD) to grow Schizosaccharomyces pombe.

Formel: C₂₀H₂Br₄Cl₄Na₂O₅
Molecular Weight: 829,6344 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00070627
CAS-Nummer: 18472-87-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0673ba109663b2e27c5b7834c380d6c9e1a3df1&vid=d81516e64843aa97474265d8bc3833093e8747b5&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR