Ferene disodium salt, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung


M-Clarity™ Qualitätsstufe = MQ200

P4272-1G P4272-25G P4272-5G
P4272-1GEA 86.7 EUR
P4272-1G P4272-25G P4272-5G
Ferene disodium salt, Sigma-Aldrich®
Ferene disodium salt

3-(2-Pyridyl)-5,6-di(2-furyl)-1,2,4-triazine-5′,5′′-disulfonic acid disodium salt, also known as Ferene, is commonly used to measure the concentration of iron ions in aqueous solutions. The reagent is sensitive to ferrous iron ions (Fe2+) and forms a purple-colored complex with these ions in the presence of a reducing agent. The intensity of the purple color formed is proportional to the concentration of iron ions present in a solution and is quantified spectrophotometrically.


  • 3-(2-Pyridyl)-5,6-di(2-furyl)-1,2,4-triazine-5′,5′′-disulfonic acid disodium salt has been used as a component in a sensitive colorimetric assay (ferene-s assay) for the detection of iron

Formel: C₁₆H₈N₄Na₂O₈S₂
Lagertemperatur: Room Temperature
MDL: MFCD00040642
CAS-Nummer: 79551-14-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e81b09379cdf3e16d606947cd1c4f103a8224da6&vid=95af8635d422f01aa072b893bdccc048e2a88969&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (colour) White to dark yellow
Appearance (form) Powder or crystals
Solubility (colour) Yellow to very dark yellow and light brown-yellow to very dark brown-yellow
Solubility (turbidity) Clear to slightly hazy
Solubility (method) Jun-03
Water ≤13.0 %
Carbon content 38.87 % (Theory), anhydrous
Nitrogen content 11.33 % (Theory), anhydrous
Infrared spectrum Conforms to structure
Bio-tests Suitable for determination of Fe
Recommended retest period 60 month


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR