Phenolrot Natriumsalz dye_SC Indicator_pH, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyme: Natriumhydrogen-4,4'-(3H-2,1-benzoxathiol-3-yliden)bisphenolat-S,S-dioxid, Phenolsulfonphthalein Natriumsalz, 4,4-(3H-2,1-Benzoxathiol-3- yliden)bisphenol-1,1-dioxid) Natriumsalz, Phenolrot wasserlöslich

MQ200

P4758-25G P4758-50G P4758-5G
P4758-25GEA 120 EUR
P4758-25G P4758-50G P4758-5G
Phenolrot Natriumsalz dye_SC Indicator_pH, Sigma-Aldrich®
Phenolrot Natriumsalz

Phenol Red sodium salt is widely used as a pH indicator. At a pH of 6,4 and below, phenol red solution will have a yellow colour. At a pH of 8,2 and above, it will have a red colour. It is commonly used in culture media to detect neutral to acidic pH changes.


  • Phenol Red sodium salt has been used as an indicator during single-guide RNA microinjection into one-cell stage zebrafish embryos
  • Suitable for use as a pH indicator

Formel: C₁₉H₁₃NaO₅S
Molecular Weight: 376.35 g/mol
Schmelzpunkt: 285 (Zers., Lit.)
Lagertemperatur: Room Temperature
MDL: MFCD00066901
CAS-Nummer: 34487-61-1
EINECS: 252-057-8
Merck Index: 12,07397
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba1dc80a4efbb86327cf195e49b5ce5bb0e2fded&vid=ccc5d83deab104d769c7ddf645398c21c3ccf71f&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) Red to Brown
Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Solubility (Colour) Dark Red
Solubility (Form), 1 mg/ml, H₂O Solution
pH Pass 6.8 (Yellow) to 8.2 (Red)
Recommended Retest Period 5 Years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR