Phenolrot Natriumsalz, Pulver, Bioreagenzien Reagenz für die Zellkultur, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyme: Natriumhydrogen-4,4'-(3H-2,1-benzoxathiol-3-yliden)bisphenolat-S,S-dioxid, Phenolsulfonphthalein Natriumsalz, 4,4-(3H-2,1-Benzoxathiol-3- yliden)bisphenol-1,1-dioxid) Natriumsalz, Phenolrot wasserlöslich

M-Clarity™ Qualitätsstufe = MQ200

P5530-25G P5530-50G P5530-5G
P5530-25GEA 185 EUR
P5530-25G P5530-50G P5530-5G
Phenolrot Natriumsalz, Pulver, Bioreagenzien Reagenz für die Zellkultur, Sigma-Aldrich®
Phenolrot Natriumsalz

Phenol Red Sodium Salt (PRSS) is a water-soluble anionic fluorescent dye, that possesses an extra Na ion attached with the SO3 group compared to phenol red. It is intended for use in cell culture applications. A solution of phenol red will have a yellow color at a pH of 6,4 or below and a red color at a pH of 8,2 and above.


The sodium salt of phenol red is used widely in culture media to identify changes from neutral to acidic pH values and as a pH marker for cell tissue culture. It is typically used in cell culture media at 11 mg/L. Phenol red in tissue culture media can act as a weak estrogen, especially in human breast cancer cells. PRSS plays a role as an indicator in the process of dying cells or overgrowth of cells in different mediums. Suitable for use as a pH indicator.

Formel: C₁₉H₁₃NaO₅S
Schmelzpunkt: 285 °C (Zers., Lit.)
Lagertemperatur: Room Temperature
MDL: MFCD00066901
CAS-Nummer: 34487-61-1
EINECS: 252-057-8
Merck Index: 12,07397
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ba92d519a36d19e08c2dad9f488a2ed031c7bfe&vid=8c9c586a0af3dfd9adff1ae99e82c9dcb82e8021&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) Red to Very Dark Red and Red-Brown and Brown-Red
Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Solubility (Turbidity), 1 mg/ml, H₂O Clear
Suitability, Suitable for use as a pH indicator Pass
Visual Interval, pH 6.8 Yellow Pass
Visual Interval, pH 7.0 Orange Pass
Visual Interval, pH 8.2 Red Pass
Cell Culture Test Pass
Insect Cell Test Pass
Recommended Retest Period 5 Years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR