Pyronin B ≥30% (Farbstoffgehalt), Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Gefahr
|
Synonyme:
3,6-Bis(diethylamino)xanthyliumchlorid
M-Clarity™ Qualitätsstufe = MQ200
P6892-25G
P6892-25GEA
442
EUR
P6892-25G
Pyronin B ≥30% (Farbstoffgehalt), Sigma-Aldrich®
Pyronin B
Pyronin B is a chromogenic reagent, and is optimal for myeloperoxidase staining, which is useful in cytomorphological diagnosis and also in leukaemia categorisation.
Formel:
C₄₂H₅₄N₄O₂·2FeCl₄ Molecular Weight: 1042,22 g/mol Schmelzpunkt: 174…176 °C Lagertemperatur: Room Temperature |
MDL:
MFCD03701356 CAS-Nummer: 2150-48-3 EINECS: 218-429-9 Merck Index: 12,08189 |
Spezifikation Testergebnisse
Appearance (Colour) | Dark Green |
Appearance (Form) | Powder |
Infrared spectrum | Conforms to Structure |
Solubility (Colour) Dark Red to Purple; c = 1mg/ml, H2O |
Conforms to Requirements |
Dye Content | ≥ 30.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...