Ferrozine mono-sodium salt hydrate Indikator, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: 5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazin-4,4''-disulfonsäure Mononatriumsalz Hydrat

M-Clarity™ Qualitätsstufe = MQ200

P9762-5G P9762-1G P9762-25G
P9762-5GEA 323 EUR
P9762-5G P9762-1G P9762-25G
Ferrozine mono-sodium salt hydrate Indikator, Sigma-Aldrich®
Ferrozine mono-sodium salt hydrate

3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-4′,4′′-disulfonic acid sodium salt is a chelating agent for ferrous iron. It is also known as Ferrozine.


  • Indicator ligand


3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-4′,4″-disulfonic acid sodium salt has been used in ferrozine chromogen solution, bioluminescence assay and in the measurement of Fe2+ concentration in the reaction medium.

Formel: C₂₀H₁₃N₄NaO₆S₂·H₂O
Schmelzpunkt: ≥300 °C
Lagertemperatur: Room Temperature
MDL: MFCD00078417
CAS-Nummer: 63451-29-6
EINECS: 274-196-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7f230a098e1ecdd28f78288edfd89a44704085b4&vid=c57ba9feff2875622691c1e75b5d48d44d60086b&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) Yellow to Yellow-Green
Appearance (Form) Powder
Solubility (Colour) Yellow to Yellow-Green
Solubility (Turbidity)
50 mg/ml, H2O
Clear to Slightly Hazy
Water (by Karl Fischer) ≤8 %
Sodium (Na) Anhydrous
By ICP-atomic emission
4.0 - 5.9 %
Suitability
For Determination of Fe++
Suitable
Recommended Retest Period 4 years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR