Venting Screw N
Lieferant: Anton Paar
104284
PAAR104284EA 0 EUR
PAAR104284
Venting Screw N
Befestigungsmaterial Schrauben
Screw for safe and convenient closing and venting of pressure vessels NXF100 and NXQ80. Consists of a PEEK body with PTFE-TFM sealing bolt.