Standard for ViscoQC Viscometers
Lieferant: Anton Paar
191118
PAAR191118EA 0 EUR
PAAR191118
Standard for ViscoQC Viscometers
Viskosimeter Rotationsviskosimeter
Silicone viscosity standard for use with ViscoQC.
Compatible with: ViscoQC 100 H, ViscoQC 100 L, ViscoQC 100 R, ViscoQC 300 H, ViscoQC 300 L, ViscoQC 300 R