Cleanroom Cubby Racks
Lieferant: Palbam
GC1302
PALBGC1302EA 12140 EUR
PALBGC1302
Cleanroom Cubby Racks
Möbel Kleiderständer
Cleanroom cubby racks.
  • Stainless steel 304L
  • Electro-polished
  • On leveler feet
  • With lip at front