Wafer casettes
Lieferant: Palbam
PCPC0101
PALBPCPC0101EA 1830 EUR
PALBPCPC0101
Wafer casettes
Wafer-Handling-Zubehör Wafer-Kassetten
The palbam class stainless steel wafer cassette is designed to replace standard plastic cassettes, in the semiconductor manufacturing process. The cassette has been designed with H Bar.
  • Non-contaminating
  • Non-deforming
  • Fully conductive
  • Withstands temperatures up to 400 ºC
  • Designed for both manual and robotic handling