Fold Down Mini Shelf
Lieferant: Palbam
WN1201
PALBWN1201EA 1090 EUR
PALBWN1201
Fold Down Mini Shelf
Möbel Regale
When not in use this mini shelf can be folded down against the wall. Ideal for tight areas where space is limited. Rounded front edge.
  • Fabrication material: Stainless steel 304 with electropolished finish