Bottle top vacuum filtration systems, AcroVac™

Lieferant: Cytiva (Formerly Pall Lab)
AVFP02S
PALLAVFP02SEA 102 EUR
PALLAVFP02S
Bottle top vacuum filtration systems, AcroVac™
Filter Flaschenaufsatzfilter
These filter units are designed for fast vacuum filtration. The low centre of gravity improves the filtration system stability and reduces the risk of system tipping during the filtration process.

  • Curved sides moulded into the reservoir
  • Finger tabs on the funnel base and storage cap allow for easy removal and tightening

PS funnel; copolyester collection reservoir; PP lid, funnel base, storage cap and vacuum port; Supor® PES membrane.

Applications: Cell culture media filtration, buffer filtration, tissue culture additive filtration, clarification of biologicals.

Systems are non-cytotoxic and meet the requirements of the Elution Test, ISO 10993-5, 1999.

Recommended/max. operating vacuum: 380/510 mm Hg
Max. temperature: 37 °C

Samples of 0,2 μm pore size systems retain a minimum of 107 cfu/cm² of Brevundimonas diminuta per modified ASTM F838; current revision.

Verpackung: Individually bagged and sterilised by gamma irradiation.
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR