Cell culture media, Dulbecco's modified eagle medium (DMEM)
Lieferant: PAN BIOTECH
Dulbecco's Modified Eagle Medium (DMEM) is a modification of Basal Medium Eagle (BME) which is Intrinsically developed for the cultivation of murine embryonic cells. DMEM is tailor-made for the cultivation of a broad range of cells, especially if the medium is supplemented with FBS.
- Suitable for most cell types
- Available in a broad variety of formulations
- Also available with stable glutamine (L-Alanyl-L-Glutamine) for a better shelf life
These media contain approximately four times as much of the vitamins and amino acids, and two to four times as much glucose depending on the version as the original formulation.
DMEM with 1,0 g/l Glucose is the standard medium, whereas DMEM with 4,5 g/l Glucose is for cells which have a high energy demand.