Cell culture media, RPMI 1640
Lieferant: PAN BIOTECH
P04-17500 P04-18000 P04-16516 P04-18047 P04-17550
PANBP04-17500EA 15.3 EUR
PANBP04-17500 PANBP04-18000 PANBP04-16516 PANBP04-18047 PANBP04-17550
Cell culture media, RPMI 1640
Zellkulturmedien
RPMI 1640 medium was developed for culture of normal and neoplastic leukocytes, but also marrow cells and hybridoma cells.
  • Serum-free media for hybridoma cells
Supplementing RPMI 1640 with varying amounts of FBS, a very good medium for many different cell lines can be obtained.