Cell culture media, Alpha MEM eagle
Lieferant: PAN BIOTECH
P04-21500
P04-21250
PANBP04-21500EA
29.1
EUR
PANBP04-21500
XXXXP04-21250
Cell culture media, Alpha MEM eagle
Zellkulturmedien
Alpha MEM is a different formulation of MEM Eagle and contains a higher concentration of amino acids. It also has a higher concentration of lipoic acid, vitamins and pyruvate. Primarily it was developed for the cultivation of hamster kidney cells, but today it is used for a broad range of mammalian cells. Among others the alpha MEM promotes the growth and progeny of bone marrow cells in suspension culture and monolayer. A further possibility is the use as a separation medium or for the out-breeding of amniotic cells.
- Sterile medium
- Store from 2 to 8 °C