Cell culture media, Hank's balanced salts (HBBS)
Lieferant: PAN BIOTECH
P04-34500
PANBP04-34500EA
21.8
EUR
PANBP04-34500
Cell culture media, Hank's balanced salts (HBBS)
Zellkulturmedien
The main purpose of buffered salt solutions is the maintenance of pH and osmolarity in cell culture applications as well as providing essential inorganic ions. These solutions are usually used for washing tissues and cells. Often they are combined with other agents in cell and tissue treatment.
- Store from 2 °C to room temperature