Cell culture media, Dulbecco's modified eagle medium/F12 Ham's (DMEM:F12)
Lieferant: PAN BIOTECH
P04-41250
PANBP04-41250EA 15.3 EUR
PANBP04-41250
Cell culture media, Dulbecco's modified eagle medium/F12 Ham's (DMEM:F12)
Zellkulturmedien
This media combines the advantages of both media DMEM (high concentration of amino acids and vitamins) and Ham’s F12 (higher concentration of zinc sulfate, putrescine and linoleic acid).
  • Suitable for clonal density cultures
  • This medium supports the growth of almost all cell lines
These are used for pancreas cells, Sertoli cells or to culture cells, which are used for human protein production.