Serum-free media, Panserin 401
Lieferant: PAN BIOTECH
P04-710401
PANBP04-710401EA
48
EUR
PANBP04-710401
Serum-free media, Panserin 401
Zellkulturmedien
Panserin 401 is a fully defined complete ready-to-use medium for the serum-free cultivation of a multitude of adherent and non adherent cells.
- Multi-purpose medium suitable for a variety of cells
- Store at 2 to 8 °C
Based on Iscove's MEM, trace elements, albumin, cholesterol, soya lipids and vitamins were added to the medium. It does not contain any growth or attachment factors.
As the medium contains no growth factors there is a possibility to investigate the effects of specific growth factors added to the cell culture.