Serum-free media, Panserin 411
Lieferant: PAN BIOTECH
Panserin 411 is a fully defined, complete ready-to-use medium for the serum-free cultivation of a multitude of adherent and non adherent cells which are Insulin-dependent (e.g. CHO-cells).
- Multi-purpose medium suitable for a variety of cells
- Store at 2 to 8 °C
In adherent as well as non adherent cells can be cultivated. As the medium contains no growth factors there is a possibility to investigate the effects of specific growth factors added to the cell culture. It does not contain any attachment factors. With some cell types a pre-treatment of the cell culture vessels with gelatine, collagen, poly-D-lysine or fibronectin may support or enable a culture under serum-free conditions.
With every adaption to serum-free media, changes of the cells should be taken into consideration. These changes may concern morphology, karyotype, surface markers and so on. Thus cells in serum-free medium may not be identical with those from cultures containing serum in which they originated (selection).
Bitte beachten For research purpose only. Not suitable for human or animal diagnostic or therapeutics procedures.